
TSKgel® SuperQ-5PW (30) Bulk Media

Synonym(s):
TSKgel® SuperQ-5PW Bulk Media
Products
Manufacturer
Physical Form
Product Line
Agency
USP Suitability
Separation Technique
Package Size
Particle Size (µm)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing