Skip to Content
MilliporeSigma
TSKgel® SuperH7000 HPLC Column phase polystyrene-divinylbenzene, L × I.D. 15 cm × 6 mm, 5 μm particle size

TSKgel® SuperH7000 (5 µm) HPLC Columns

TSKgel® SuperH7000 HPLC Column phase polystyrene-divinylbenzene, L × I.D. 15 cm × 6 mm, 5 μm particle size
Synonym(s):
TSKgel® SuperH Columns

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1