
Tetramethyl Ethylene Diamine

Synonym(s):
N,N,N′,N′-Tetramethylethylenediamine, 1,2-Bis(dimethylamino)ethane, TEMED, TMEDA
Linear Formula:
(CH3)2NCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
116.20
Products
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Agency
Format
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing