
Soy Peptone for Biotechnology and Microbiology

Synonym(s):
Peptone from soybean
CAS Number:
EC Number:
Products
Product Category
Biological Source
Color
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing