Skip to Content
MilliporeSigma
Sodium bis(trifluoromethylsulfonyl)imide ≥99.5% trace metals basis, anhydrous, battery grade

Sodium bis(trifluoromethylsulfonyl)imide

Sodium bis(trifluoromethylsulfonyl)imide ≥99.5% trace metals basis, anhydrous, battery grade
Synonym(s):
Sodium trifluoromethanesulfonimide, Sodium bis(trifluoromethylsulfonyl)imide
Empirical Formula (Hill Notation):
C2F6NNaO4S2
CAS Number:
Molecular Weight:
303.14

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1