
Purospher® STAR RP-18 Endcapped (2 µm) Capillary Column

Synonym(s):
Purospher® STAR RP-18 Endcapped (2 µm) Capillary Columns
Products
Brand
Agency
USP Suitability
Separation Technique
ID (mm)
Length (mm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing