
N-Nitrosodibutylamine

Synonym(s):
N-Nitrosodibutylamine, Dibutyl nitrosamine, NDBA
Empirical Formula (Hill Notation):
C8H18N2O
CAS Number:
Molecular Weight:
158.24
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing