
N-Desethylamiodarone hydrochloride

Synonym(s):
(2-Butylbenzofuran-3-yl)[4-[2-(ethylamino)ethoxy]-3,5-diiodophenyl]methanone hydrochloride
Empirical Formula (Hill Notation):
C23H25I2NO3·HCl
CAS Number:
Molecular Weight:
653.72
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing