Skip to Content
MilliporeSigma
Peptone, mycological suitable for microbiology

Mycological Peptone for Biotechnology and Microbiology

Peptone, mycological suitable for microbiology
Synonym(s):
Peptone from bovine meat
CAS Number:

Products

Product Category
Biological Source
pH Value
Physical Form
Tissue
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1