Skip to Content
CA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
Malt Extract for Biotechnology and Microbiology
Synonym(s):
Malt extract
CAS Number:
8002-48-0
Products
Product Category
extracts for media (2)
Brand
Millipore (1)
Biological Source
Color
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Sterility
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.05391
Malt Extract for Biotechnology and Microbiology,
Prime, for Biotechnology and Microbiology
Expand
View Pricing
70167
Malt Extract for Biotechnology and Microbiology,
for Biotechnology and Microbiology
Expand
View Pricing