
Intermediates for Further Manufacturing Use (FFMU) - RHD (Rh) Specificities

Products
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing