Skip to Content
MilliporeSigma
Hexyl 2-methylbutanoate ≥95%, FCC, FG

Hexyl 2-methylbutanoate

Hexyl 2-methylbutanoate ≥95%, FCC, FG
Synonym(s):
Hexyl 2-methylbutyrate
Linear Formula:
C2H5CH(CH3)CO2(CH2)5CH3
CAS Number:
Molecular Weight:
186.29
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing