Skip to Content
MilliporeSigma
Fluid K - Ready-to-use Rinse Fluid bottle capacity 125 mL, bottle filling volume 100 mL, closure type, Black screw cap with septum and protector, pack of 12 bottles

Fluid K - Ready-to-use Rinse Fluid

Fluid K - Ready-to-use Rinse Fluid bottle capacity 125 mL, bottle filling volume 100 mL, closure type, Black screw cap with septum and protector, pack of 12 bottles

Products

Product Category
Brand
Physical Form
Agency
Capacity (mL)
Quality Segment
Closure Type
Application
Package Size
Sterility
Sterilization Method