
Desethylamiodarone

Synonym(s):
(2-Butylbenzofuran-3-yl)[4-[2-(ethylamino)ethoxy]-3,5-diiodophenyl]methanone hydrochloride
Empirical Formula (Hill Notation):
C23H25I2NO3·HCl
CAS Number:
Molecular Weight:
653.72
Products
Feature
Formula Weight
Manufacturer
Agency
Format
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing