Skip to Content
MilliporeSigma
vibrant-m

(9β,11β,16α)-21-Chloro-9,11-epoxy-17-[(2-furanylcarbonyl)oxy]-16-methylpregna-1,4-diene-3,20-dione

vibrant-m
Empirical Formula (Hill Notation):
C27H29ClO6
CAS Number:
Molecular Weight:
484.97

Products

Feature
Formula Weight
Quality Segment
Application
Package Size
API Family
Special Grade