
2H,2H,3H,3H-Perfluorooctanoic acid

Synonym(s):
4,4,5,5,6,6,7,7,8,8,8-Undecafluorooctanoic acid
Empirical Formula (Hill Notation):
C8H5F11O2
CAS Number:
Molecular Weight:
342.11
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing