MilliporeSigma
Acids
Acids bottles
In Life Science, from complex analysis to routine lab work or production, each demand is unique that requires distinct solution and is governed by different regulations. When looking for products, you have to consider the targeted applications and your specific needs.
To simplify your search, our extensive range of acids is offered under three portfolio brands intended for different applications: Supelco® products for analytical chemistry, Sigma-Aldrich® materials for lab and production, SAFC® products for biopharmaceutical and pharmaceutical development and manufacturing. Each brand is offered in a variety of volumes, packaging materials, and with different documentation packages. Now, explore and discover the wide range of product portfolio that suits your application.
Suprapur® Hydrofluoric acid 40% from Supelco®
High Purity Acids for Instrumental Analysis
We offer a comprehensive range of LiChropur® high purity acids for HPLC and LC-MS analysis to minimize background noise and signal suppression. These acids are application-tested by our quality control to meet our customers’ expectations.
Our range of Suprapur® and Ultrapur® products are designed to accommodate your need to buy high purity acids for inorganic trace and ultratrace analysis. Whether you need to meet international standards, follow safety regulations, require large or small quantities you are sure to find the ideal solution in this product range. The products and quality control facilities always comply with the same guidelines and regulations as yours, so quality assurance is of prime importance. We offer tailored range of high purity acids to satisfy the demand for sensitive instrumental techniques (ICP-OES, ICP-MS, AAS, etc.).
Acids for Classical Analysis
EMSURE® acids for analysis are our premium grade for all regulated and highly demanding lab applications. Most EMSURE® acids for analysis meet or exceed the requirements of ACS, ISO and/or Reag. Ph Eur. They feature the best and most extensive product specifications available worldwide as well as the widest range of pack sizes and packaging options – from 25 mL glass bottles up to 200 L PE barrels.
If you need less performance, but still top-notch quality and reliability, our acids for analysis EMPARTA® are for you. They meet the requirements of ACS and are suitable for a multitude of analytical lab applications. Additionally, our EMPLURA® grade of acids covers the most important specification parameters and is suitable for preparative lab work, cleaning and production.
For maximum safety in your lab operations, we have developed innovative packaging solutions like our Safebreak bottle, a PE-coated glass bottle, and the SafetyCap for reagents generating excess pressure in the bottle. With our acids for analysis, you benefit from application-oriented quality, more flexibility and safety, and an excellent cost-performance ratio.
Biopharmaceutical and Pharmaceutical Formulation and Production Products
In pharmaceutics, the regulatory world is everchanging. Keeping up with the latest requirements, while not losing track of the market itself is definitely not an easy task to fulfill. And as the pressure caused by generics and biosimilars making their way into the market keeps rising, it becomes more and more important to focus on what really matters: Quality and the right partner throughout your product journey.
We support you in every step of development, scale-up, and production with services that range from process optimization to regulatory affairs support. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
As part of our Emprove® Program, our raw materials also come with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation.
Volumetric Solutions for Titration
Our Titripur® volumetric solutions and Titrisol® concentrates offer a range of acids in different concentrations for quantifying compounds with the titration method. They are produced and analyzed under high quality standards. The Titripur® solutions are certified according to ISO/IEC 17025. An innovation in this area are our digital products with an RFID tag on the bottles for seamless data transfer.