Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Form:
coarse powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

≥99.8% (calc. to the dried substance)

form

coarse powder

impurities

residual solvents, complies
≤0.001% total nitrogen (N)
≤0.003% total S (as SO4)
≤0.01% in water insoluble matters

loss

≤0.5% loss on drying, 300 °C

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
silicate (as SiO2): ≤20 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤10 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤20 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤20 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carbonate (Soda) is a white salt. It can be synthesized by the electrolysis of molten sodium hydroxide or sodium chloride.

Application

Sodium carbonate may be used for the determination of total polyphenolic index of wine products from grape by the Folin-Ciocalteu method. It has been used to compose the ELISA (Enzyme-Linked Immunosorbent Assay) coating buffer. It may be used to prepare the antibody coating buffer for use in IGF-I kinase receptor activation assay (KIRA) assay.

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service