Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C84
CAS Number:
Molecular Weight:
1008.90
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

98%

form

powder

solubility

organic solvents: soluble

SMILES string

C1CC2CCC3CC4CC5c6cc7cc8cc9CC%10CCC%11CC1C%12c%13c%11c%10c%14c9c%15c8c%16c7c6c%17C%18=C5C4c%19c3c%20c2c%12c%21c%20c(c%18%19)c%22c%17c%16c%15c%23c%14c%13c%21c%22%23

Features and Benefits

Material is most likely a mixture of the D2d and D2 structural isomers. Most common fullerene impurities are C82 and C86.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service