Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium dioxide, anatase
Synonym(s):
Titanium dioxide
Linear Formula:
TiO
2
CAS Number:
1317-70-0
Molecular Weight:
79.87
EC Number:
215-280-1
Products
Product Category
oxides (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Particle Size (µm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
637254
Titanium dioxide, anatase,
nanopowder, <25 nm particle size, 99.7% trace metals basis
Expand
View Pricing
232033
Titanium dioxide, anatase,
powder, 99.8% trace metals basis
Expand
View Pricing
248576
Titanium dioxide, anatase,
powder, −325 mesh, ≥99% trace metals basis
Expand
View Pricing