Introduction to Microbial Media
- Introduction
- Types of microbial media
- FAQs
- Microbial medial selection guide
Introduction
Microbiological studies require the availability of different species of bacteria, yeast or viruses in the laboratory. Large scale processes such as fermentation, protein and vaccine production require large numbers of bacteria in physiologically active state. A suitable nutrient medium that provides appropriate biochemical environment and maintains all the characteristics of microbes is therefore required for various applications.
Any microbial medium should include sources of nutrition, energy, essential minerals, buffering agents and pH indicators. Sometimes they also include selective agents and solidifying agents. Obligate parasites such as bacteriophages may be cultivated in a nutrient medium that also contains host bacteria.
The following are the essential components of a typical microbial medium:
- Nutrition sources – sources of proteins, vitamins, mineral and carbohydrates; mostly mixture of peptone and meat extract
- Energy sources – sources of carbohydrates; mostly glucose
- Essential minerals – sources of micro and macro minerals; mostly present in peptone and meat extract
- Buffering agents – agents that maintain optimum pH; mostly specific amino acids, phosphates, citrates and zwitterions
- pH indicators – agents that indicate variation in pH of the medium by change in color; mostly phenol red
- Selective agents – agents that allow the growth of only specific bacteria; antibiotics, tellurites, azides and bile salts are examples
- Solidifying agents – agents that maintain the medium in solid or semi-solid state, if required; widely used agent is agar though gelatin is sometimes used
- Specific substances such as growth factors, enzymes may be incorporated into the medium for specific bacteria
Types of microbial media
- Simple or basal media: Include nutrient broth and peptone water; used routinely to isolate and culture a variety of bacteria in a molecular biology research laboratory
- Complex media: Contain mixture of a variety of nutrients; the exact composition of amino acid source is not defined. Examples include chocolate agar, MacConkey agar, Lowenstein-Jensen medium
- Defined media: Contain known and defined sources of carbon and nitrogen as required by certain bacteria
- Special media: Contain specific substances that encourage growth of fastidious bacteria, discourage the growth of unwanted bacteria or contain indicators that distinguish different types of bacteria
Name | Introduction | Application |
Bacterial Media | ||
LB media | Lysogeny Broth or LB is the most popular and commonly used medium to culture E. coli. It was first created by Bertani G to culture Shigella indicator strain. LB is a rich medium containing peptone, yeast extract, NaCl and agar (for solid medium). Depending on the concentration of NaCl, the LB medium may be classified as follows: LB - Miller, LB - Lennox and LB - low salt. | LB media are suitable for non-selective cultivation of E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. Plates containing sterile LB agar with or without antibiotics are also available for selective culture of antibiotic-resistant strains of E. coli. |
Minimal salts (M9) | M9 medium contains essential salts and nitrogen. | Minimal salts (M9) medium is suitable for non-selective cultivation of E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. |
Terrific Broth - modified | Terrific broth is a nutrient rich medium that results in higher yield of bacteria in less time. | Terrific broth (modified) is suitable for non-selective cultivation of E. coli strains for cloning and high yield production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. |
Hanahan’s Broth (SOB Medium) | SOB medium has higher peptone, amino acids and peptides than LB medium. | Hanahan’s Broth (SOB medium) enables the preparation of high efficiency competent cells. |
SOC Medium | SOC medium is a modified SOB medium with added glucose. In the presence of glucose the cellular processes utilize other sugars are repressed. The expression of recombinant proteins that might affect the host cell physiology is inhibited thus resulting in high efficiency transformation reactions. | SOC medium enables the high efficiency transformation of competent cells. |
2X YT medium | 2X YT medium contains double the concentration of yeast extract compared to typical LB medium. It is useful for the growth and maintenance of high density of cells for a longer growth period. | 2X YT medium is suitable for non-selective cultivation of M13 bacteriophage and E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. |
NZCYM Broth | NZCYM Broth is nutrient rich broth with amino acids, peptides, vitamins and low salt concentration. | NZCYM Broth is suitable for non-selective cultivation of lambda bacteriophage and E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. |
Yeast Media | ||
Yeast Nitrogen Base with or without amino acids | Yeast Nitrogen Base is nutrient-rich medium with sources of nitrogen, vitamins, trace elements and salts. It acts as the basic nutrient medium for most fermentation reactions involving yeast cultures. | Yeast Nitrogen Base with and without added amino acids for the cultivation of yeast and for classification based on amino acid and carbon requirements, respectively. |
Phage Media | ||
NZ Amine®Broth | NZ Amine® Broth is nutrient rich nutrient-rich medium with amino acids, peptides, vitamins and low salt concentration. | NZ Amine® Broth is suitable for the growth of lambda bacteriophage. |
FAQs
What are the differences among the Luria, Lennox and Miller LB formulations?
LB, (originally termed lysogeny broth) was initially composed of tryptone, yeast extract, NaCl and glucose.Soon after, the glucose was omitted (Miller's version), and later the NaCl content lowered by half (Lennox's version).For some applications, even lower salt is required (Luria's low salt version).
What is the difference among the LB - Miller products?
LB - Miller is available in many types to suit your needs.The different product formats include powder and liquid form.The powder form is also available with agar for easy LB-agar plate preparation.L2542 (LB Miller liquid)L3522 (LB Miller powder)L3147 (LB Miller powder with agar)
Which bacterial culture medium is the best choice for my application?
Each of the broths will likely grow E. coli very well, but there are still general guidelines for choosing a broth when you are working without a protocol.Generally:LB - Miller and LB - Lennox are used for E. coli growth and maintanence, DNA plasmid production and protein production.The Lennox formulation has a lower salt content required for some salt-sensitive selection antibiotics.LB - Luria low salt is used for special applications where the E. coli growth or other constraints require the lowest possible salt content.Terrific Broth is used for higher yield protein production and high yield DNA plasmid production, because of the faster growth of the E. coli in this medium.SOB is used for protein production, DNA plasmid production and the generation of high-efficiency competent cells.SOC is used for initial growth of competent cells and the transformation procedure.
Will adding magnesium to the culture medium increase cell density?
In microbial broth formulations that do not already contain magnesium, the addition of 10-20 mM MgCl2 or MgSO4 may increase cell densities. You may need to also increase the shaking speed of the incubator.
Name | Components | Product No. | Format | Preparation Notes | Application |
LB – Miller | • 10 g/L Tryptone • 5 g/L yeast extract • 10 g/L NaCl | L2542 | Liquid | Ready-to-use | Suitable for regular E. coli culture. |
•10g/L Tryptone •10g/L NaCl •5g/L Yeast Extract | L3522 | Powder | Mix 25 g in 1L water; autoclave and cool before use | ||
•15g/L Agar •10g/L Tryptone •10g/L NaCl •5g/L Yeast Extract | L3147 | Powder with agar | Mix 40 g in 1L water, heat to dissolve; autoclave and cool before use | ||
•15g/L Agar •10g/L Tryptone •10g/L NaCl •5g/L Yeast Extract •100μg/ml ampicillin | L5667 | Agar plates with 100 μg/ml ampicillin | Ready-to-use | ||
•15g/L Agar •10g/L Tryptone •10g/L NaCl •5g/L Yeast Extract •50μg/ml ampicillin | L0168 | Agar Plate with 50 μg/ml ampicillin | |||
•15g/L Agar •10g/L Tryptone •10g/L NaCl •5g/L Yeast Extract •50μg/ml kanamycin | L0543 | Agar plates with 50 ug/ml kanamycin | |||
•15 g/L Agar •10 g/L Tryptone •10 g/L NaCl •5 g/L Yeast Extract •100 μg/ml carbenicillin | L0418 | Agar plates with 100 μg/ml carbenicillin | |||
LB – Lennox | • 10 g/L peptone • 5 g/L yeast extract • 10 g/L NaCl | L3022 | Powder | Mix 20 g in 1L water; autoclave and cool before use | Suitable for culture of E. coli strains that are salt-sensitive. |
•15 g/L Agar •10 g/L Tryptone •5 g/L Yeast Extract •5 g/L NaCl | L2897 | Powder with agar | Mix 35 g in 1L water, heat to dissolve; autoclave and cool before use | ||
•10 g/L Tryptone •5 g/L Yeast Extract •5 g/L NaCl •2.2 g/L inert binding agents | L7275 | Tablets | Add each 1.2 g tablet to 48.3 mL water; autoclave and cool before use | ||
•13.72g/L Agar •9.14g/L Tryptone •4.57 g/L Yeast Extract •4.57 g/L NaCl •1.6g/L inert binding agents | L7025 | Tablets with agar | Add each 1.68 g tablet to 48.3 mL water; autoclave and cool before use | ||
•10g/L Tryptone •5 g/L Yeast Extract •5 g/L NaCl •0.6 g/L inert binder (EZMix formulation only) | L7658 | EZMix™ | Mix 20.6 g in 1L water; autoclave and cool before use | ||
•15g/L Agar •10g/L Tryptone •5 g/L Yeast Extract •5 g/L NaCl | L7533 | EZMix™ with agar | Mix 35.6 g in 1L water, heat to dissolve; autoclave and cool before use | ||
•15g/L Agar •10g/L Tryptone •5 g/L Yeast Extract •5 g/L NaCl | L5542 | Plates | Ready-to-use | ||
•10 g/L Tryptone •5.0 g/L Yeast Extract •5.0 g/L NaCl | L9234 | Liquid | Ready-to-use | ||
LB – low salt | • 10 g/L peptone • 5 g/L yeast extract • 0.5 g/L NaCl (and 15 g/L agar for solid medium) | L3397 | Powder | Mix 15.5 g in 1L water; autoclave and cool before use | Suitable for culture of E. coli strains that require low salt conditions to grow. |
•15g/L Agar •10g/L Tryptone •5g/L Yeast Extract •0.5g/L NaCl | L3272 | Powder with agar | Mix 30.5 g in 1L water, heat to dissolve; autoclave and cool before use | ||
Overnight Express Instant LB Medium | • Overnight Express Autoinduction System 1 components • Luria Bertani (LB) broth | 71757-M | Granulated medium | Just add the EasyPak contents to 1 L sterile water, supplement with 10 ml glycerol, and microwave for 2 min to dissolve. Overnight Express Instant LB Medium is also available in 1 kg bottles. Granulated medium ensures rapid, uniform dissolution in water, prevents clumping of the medium, and avoids inhalation of airborne powder. | Enables regulated protein expression in E. coli, without monitoring the culture or adding IPTG during cell growth. |
Overnight Express Instant TB Medium | • Overnight Express Autoinduction System 1 components • Terrific Broth (TB) medium | 71491 | Granulated medium | EasyPak is an aluminum foil pouch containing 60 g granulated medium, sufficient for 1 L culture. Just add the EasyPak contents to 1 L sterile water, supplement with 10 ml glycerol, and microwave for 2 min to dissolve. Overnight Express Instant TB Medium is also available in 1 kg bottles. Granulated medium ensures rapid, uniform dissolution in water, prevents clumping of the medium, and avoids inhalation of airborne powder. |
Name | Components | Product No. | Format | Preparation Notes | Application |
Minimal salts (M9) | • 33.9 g/L Na2HPO4.7H2O •15 g/L KH2PO4 • 5 g/L NH4Cl and 2.5 g/L NaCl | M6030 | Powder | Mix 56.4 g in 1L water to obtain 5X stock solution; autoclave, cool and dilute to 1X before use | Suitable for recombinant E. coli strains |
•15 g/L KH2PO4 •64 g/L Na2HPO4 •2.5 g/L NaCl •5.0 g/L NH4Cl | M9956 | Liquid | Ready-to-use |
Name | Components | Product No. | Format | Preparation Notes | Application |
Terrific Broth - modified | • 23.6 g/L yeast extract • 11.8 g/L tryptone • 9.4 g/L K2HPO4 • 2.2 g/L KH2PO4 | T5574 | Liquid | Ready-to-use | Suitable for non-selective cultivation of E. coli strains for cloning and high yield production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. |
T9179 | EZMix™ | Mix 48.2 g in 1L water, add 4 mL glycerol; autoclave and cool before use |
Name | Components | Product No. | Format | Preparation Notes | Application |
2X YT medium | • 16 g/L tryptone • 10 g/L yeast extract • 5 g/L NaCl | Y1003 | Liquid | Ready-to-use | Suitable for non-selective cultivation of M13 bacteriophage and E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. |
Y2377 | Powder | Mix 31 g in 1L water; autoclave and cool before use | |||
Y2627 | EZMix™ | Mix 31.6 g in 1L water; autoclave and cool before use |
Name | Components | Product No. | Format | Preparation Notes | Application |
NZCYM Broth | • 10 g/L tryptone • 5 g/L yeast extract • 5 g/L NaCl • 1 g/L Casamino acids • 1 g/L MgSO4 | N3643 | Powder | Mix 22 g in 1L water; autoclave and cool before use | Suitable for non-selective cultivation of lambda bacteriophage and E. coli strains for cloning, production of DNA, plasmid DNA and recombinant proteins. It is also suitable for selective cultivation when appropriate antibiotics are added. |
Name | Components | Product No. | Format | Preparation Notes | Application |
Yeast Nitrogen Base with amino acids | Nitrogen Sources: •Ammonium sulfate, 5.0 g/L Amino Acids: •L-histidine, 10 mg/L •DL-methionine, 20 mg/L •DL-tryptophan, 20 mg/L Vitamins: •Biotin, 2.0 •Calcium pantothenate, 400 •Folic acid, 2.0 •Inositol, 2.0 mg/L •Nicotinic acid, 400 •p-Aminobenzoic acid, 200 •Pyridoxine HCl, 400 •Riboflavin, 200 •Thiamine HCL, 400 Trace Elements: •Boric acid •Copper sulfate •Potassium iodide •Ferric chloride •Magnesium sulfate •Sodium molybdate •Zinc sulfate Salts: •Potassium phosphate monobasic, 1.0 g/L •Magnesium sulfate, 0.5 g/L •Sodium chloride, 0.1 g/L •Calcium chloride, 0.1 g/L | Y1250 | Powder | Mix 6.7 g in 100 mL water, add 5 g glucose to obtain 10X stock solution; filter sterilize and dilute to 1X with sterile water before use | Yeast Nitrogen Base is a highly-referenced growth medium used for the cultivation of yeast. This nutrient-rich microbial broth contains amino acids, nitrogen, vitamins, trace elements and salts. |
Yeast Nitrogen Base without amino acids | Nitrogen Sources: •Ammonium sulfate, 5.0 g/L Vitamins: •Biotin, 2.0 •Calcium pantothenate, 400 •Folic acid, 2.0 •Inositol, 2.0 mg/L •Nicotinic acid, 400 •p-Aminobenzoic acid, 200 •Pyridoxine HCl, 400 •Riboflavin, 200 •Thiamine HCL, 400 Trace Elements: •Boric acid, 500 •Copper sulfate, 40 •Potassium iodide, 100 •Ferric chloride, 200 •Manganese sulfate, 400 •Sodium molybdate, 200 •Zinc sulfate, 400 Salts: •Potassium phosphate •monobasic, 1.0 g/L •Magnesium sulfate, 0.5 g/L •Sodium chloride, 0.1 g/L •Calcium chloride, 0.1 g/L | Y0626 | Powder | Mix 6.8 g in 100 mL water, add 5 g glucose and 5-10 mg of appropriate amino acids to obtain 10X stock solution; filter sterilize and dilute to 1X with sterile water before use | Yeast Nitrogen Base is considered ideal for culturing wild type yeast. The presence of amino acid is known to prevent selectable marker utilization. Hence, this media is formulated without amino acids and is useful for cloning and manipulation of yeast artificial chromosome. |
Name | Components | Product No. | Format | Preparation Notes | Application |
NZ Amine®Broth | • 10 g/L tryptone • 5 g/L NaCl • 1 g/L MgSO4 | N3518 | Powder | Mix 16 g in 1L water; autoclave and cool before use | Suitable for the growth of lambda bacteriophage. |