Merck
All Photos(1)
Key Documents
MAK490
Sigma-Aldrich
L-Amino Acid Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Quality Level
application(s)
food and beverages
detection method
colorimetric
fluorometric
relevant disease(s)
genitourinary disorders; cancer
shipped in
wet ice
storage temp.
−20°C
General description
L-amino acids are essential molecules defined by the nature of their side-chain. There are 20 amino acids that are commonly used by most organisms for the production of proteins and other biologically relevant molecules. While amino acids can occur in either the D- or L-enantimer, only the L-form is used by cells.
Features and Benefits
  • Linear detection range: 3.3 to 500 µM (colorimetric assay) and 0.13 to 50 µM (fluorometric assay) for 60 minute reaction.
  • Compatible with high-throughput handling systems.
Suitability
The kit is suitable for L-Amino Acid determination in serum, culture media, tissue homogenates, cell lysates, urine, food samples, etc.
Principle
The L-Amino Acid Assay Kit uses an enzyme-catalyzed oxidation of L-amino acids to convert a dye into a colored or fluorescent form. The absorbance at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 is directly proportional to the L-amino acid concentration in the sample.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service