Merck
HomeTitration & Karl Fischer TitrationDetermination of Water Content in Sodium Chloride Using Karl Fischer Titration
Determination of Water Content in Sodium Chloride Using Karl Fischer Titration
Product Group
Inorganic compounds
General Information concerning the product group
Inorganic compounds
Salts contain water in different kinds of bonding states: adherent moisture - enclosed water - crystal water. For determination of the total water content the substance must be completely dissolved. If the salt does not dissolve in methanol sufficiently, formamide can be added to the recipient vessel and/or titration carried out at 50 °C. If adherent moisture alone is of interest the dissolution of the sample must be prevented through addition of a non-polar solvent (e.g. chloroform) to the KF solvent. A good alternative for salts to the direct volumetric determination is often the Karl Fischer oven technique. Through slow heating of the sample it is hereby possible to differentiate among the various types of water bonding. For salts that cause disturbing side reactions in the KF solutions (e.g. carbonates, sulphites, borates) the KF oven technique is the method of choice. The same is also true for oxides and hydroxides. These alkaline compounds react with the weakly acidic KF solution resulting in the formation of water, which is erroneously detected as water during the titration. Aqueous inorganic acids and bases are titrated volumetrically. Since stoichiometry and reaction rate of the Karl Fischer reaction are influenced by the pH, sufficient buffering of the KF solution is a prerequisite for reproducible and accurate results. Optimum conditions are obtained in the neutral pH range (pH 4 - 8). Thus, the determination of acids is performed through the addition of Aquastar® buffer solution for strong acids, imidazole or a derivative of imidazole to the titration media. For the titration of bases the pH is adjusted by addition of Aquastar® buffer solution for strong bases, salicylic acid or benzoic acid. With particularly strong, highly concentrated acids the propensity for esterification in methanolic solutions should also be noted. In such cases external neutralisation in a methanol-free medium is necessary.
Special Information concerning the sample and the methods
Alkali halides do not contain any constitutional water but very firmly bonded, enclosed water, which can only be determined through complete dissolution of the sample. For direct volumetric analysis in methanolic KF solvents the addition of formamide promotes the dissolution. Due to the low concentration of water a titrant with a factor of 2 mg/mL is recommended. The employment of KF oven technology in combination with coulometry is also possible for determination of adherent water. The enclosed water is often only very slowly released even at higher temperatures.
Titration One Component System
Reagents:
Titrant
188002 Aquastar® - CombiTitrant 2 - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 2 mg water
Solvent
30 mL 188009 Aquastar® - CombiMethanol - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
and
20 mL 109684 Formamide - as solubiliser
Titration Parameters:
Extraction time: 3 min.
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
1 - 3 g
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. An extraction time of three minutes is recommended.
Titration Two Component System
Reagents:
Titrant
188011 Aquastar® - Titrant 2 - Titrant for volumetric titration with two component reagents, 1 mL = approx. 2 mg water
Solvent
30 mL 188015 Aquastar® - Solvent - Solvent for volumetric titration with two component reagents
and
20 mL 109684 Formamide - as solubiliser
Titration Parameters:
Stirring time: 3 min.
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample size:
1 - 3 g
Procedure:
The titration medium is first placed into the cell and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. An extraction time of three minutes is recommended.