Merck
HPLC Analysis of Paraben Preservatives on Discovery® C18
HPLC Analysis of Paraben Preservatives on Discovery® C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® C18 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] acetonitrile: [B] water, (30:70, A:B)
flow rate
1 mL/min
column temp.
20 °C
detector
UV, 254 nm
injection
10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany