Merck
HPLC Analysis of Monosaccharides Ribose, Xylitol, and Mannose on Chromolith® NH2
HPLC Analysis of Monosaccharides Ribose, Xylitol, and Mannose on Chromolith® NH2 application for HPLC
Materials
standard
Product No.
Description
Pricing
Xylitol
analytical standard
Mannose
United States Pharmacopeia (USP) Reference Standard
Ribose
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
Chromolith NH2, 10 cm x 4.6 mm I.D., 2 μm particles (1.52028.0001)
mobile phase
[A] acetonitrile; [B] water; (85:15, A:B)
flow rate
3 mL/min
pressure
47 bar (682 psi)
column temp.
23 °C
detector
UV, 190 nm
injection
5 μL
sample
ribose 5.9 mg/mL, xylitol 8.1 mg/mL, mannose 7.6 mg/mL in mobile phase
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany