GC Analysis of Glycerides in Lard on MET-Biodiesel
CONDITIONS
column
MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 μm with integrated 2 m x 0.53 mm I.D. guard (28668-U)
oven
150 °C, 20 °C/min to 350 °C (15 min)
detector
FID, 400 °C
carrier gas
helium, 15 mL/min
injection
1 μL, cool on-column
sample
lard extract