Merck
GC Analysis of an 8-Component Solvent Mix in Water on Watercol 1910
GC Analysis of an 8-Component Solvent Mix in Water on Watercol™ 1910 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Watercol 1910 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
Watercol 1910, 30 m x 0.25 mm I.D., 0.20 μm (29711-U)
oven
35 °C, 4 °C/min to 125 °C (2 min)
inj. temp.
250 °C
detector
TCD, 300 °C
carrier gas
helium, 25 cm/sec
injection
1 μL, 100:1 split
liner
4 mm I.D., split type, cup design
sample
8-component solvent mix in water
Description
Analysis Note
The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water does not interfere chromatographically with other small polar analytes. This application shows the analysis of an 8-component solvent mix in water on Watercol 1910.
Legal Information
Watercol is a trademark of Sigma-Aldrich Co. LLC