Merck
  • Subcellular Localization and Activity of TRPM4 in Medial Prefrontal Cortex Layer 2/3.
Subcellular Localization and Activity of TRPM4 in Medial Prefrontal Cortex Layer 2/3.
Frontiers in cellular neuroscience (2018-02-15)
Denise Riquelme, Ian Silva, Ashleigh M Philp, Juan P Huidobro-Toro, Oscar Cerda, James S Trimmer, Elias Leiva-Salcedo
ABSTRACT
TRPM4 is a Ca
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Nystatin, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Goat serum