Merck
  • Redox regulation of PEP activity during seedling establishment in Arabidopsis thaliana.
Redox regulation of PEP activity during seedling establishment in Arabidopsis thaliana.
Nature communications (2018-01-05)
Manuel Guinea Díaz, Tamara Hernández-Verdeja, Dmitry Kremnev, Tim Crawford, Carole Dubreuil, Åsa Strand
ABSTRACT
Activation of the plastid-encoded RNA polymerase is tightly controlled and involves a network of phosphorylation and, as yet unidentified, thiol-mediated events. Here, we characterize PLASTID REDOX INSENSITIVE2, a redox-regulated protein required for full PEP-driven transcription. PRIN2 dimers can be reduced into the active monomeric form by thioredoxins through reduction of a disulfide bond. Exposure to light increases the ratio between the monomeric and dimeric forms of PRIN2. Complementation of prin2-2 with different PRIN2 protein variants demonstrates that the monomer is required for light-activated PEP-dependent transcription and that expression of the nuclear-encoded photosynthesis genes is linked to the activity of PEP. Activation of PEP during chloroplast development likely is the source of a retrograde signal that promotes nuclear LHCB expression. Thus, regulation of PRIN2 is the thiol-mediated mechanism required for full PEP activity, with PRIN2 monomerization via reduction by TRXs providing a mechanistic link between photosynthetic electron transport and activation of photosynthetic gene expression.
Sigma-Aldrich
Anti-Chicken IgY (IgG) (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phytagel, BioReagent, suitable for plant cell culture, powder