Merck
  • Intrinsic Ribosome Destabilization Underlies Translation and Provides an Organism with a Strategy of Environmental Sensing.
Intrinsic Ribosome Destabilization Underlies Translation and Provides an Organism with a Strategy of Environmental Sensing.
Molecular cell (2017-11-04)
Yuhei Chadani, Tatsuya Niwa, Takashi Izumi, Nobuyuki Sugata, Asuteka Nagao, Tsutomu Suzuki, Shinobu Chiba, Koreaki Ito, Hideki Taguchi
ABSTRACT
Nascent polypeptides can modulate the polypeptide elongation speed on the ribosome. Here, we show that nascent chains can even destabilize the translating Escherichia coli ribosome from within. This phenomenon, termed intrinsic ribosome destabilization (IRD), occurs in response to a special amino acid sequence of the nascent chain, without involving the release or the recycling factors. Typically, a consecutive array of acidic residues and those intermitted by alternating prolines induce IRD. The ribosomal protein bL31, which bridges the two subunits, counteracts IRD, such that only strong destabilizing sequences abort translation in living cells. We found that MgtL, the leader peptide of a Mg
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
N-Formyl-L-methionine, ≥90% (TLC)
Roche
High Pure miRNA Isolation Kit, kit of for 50 isolations