Merck
  • Endogenous synthesis of milk oleic acid in dairy ewes: In vivo measurement using
Endogenous synthesis of milk oleic acid in dairy ewes: In vivo measurement using
Journal of dairy science (2017-05-22)
P G Toral, P Frutos, D Carreño, G Hervás
ABSTRACT
The use of stable isotopes is a reliable and risk-free alternative to radioactive tracers for directly examining in vivo fatty acid (FA) metabolism. However, very limited information is available in ruminants, and none is available in sheep. Therefore, we conducted an experiment in dairy ewes to determine, for the first time in this species, the uptake, Δ
Sigma-Aldrich
Linoleic acid methyl ester, cis/trans-isomers
Sigma-Aldrich
Linoleic acid, conjugated methyl ester
Supelco
F.A.M.E. Mix, C4-C24, certified reference material,  wt. % (varied)
Sigma-Aldrich
Linolenic acid methyl ester isomer mix