Merck
  • Ceapins are a new class of unfolded protein response inhibitors, selectively targeting the ATF6α branch.
Ceapins are a new class of unfolded protein response inhibitors, selectively targeting the ATF6α branch.
eLife (2016-07-21)
Ciara M Gallagher, Carolina Garri, Erica L Cain, Kenny Kean-Hooi Ang, Christopher G Wilson, Steven Chen, Brian R Hearn, Priyadarshini Jaishankar, Andres Aranda-Diaz, Michelle R Arkin, Adam R Renslo, Peter Walter
ABSTRACT
The membrane-bound transcription factor ATF6α plays a cytoprotective role in the unfolded protein response (UPR), required for cells to survive ER stress. Activation of ATF6α promotes cell survival in cancer models. We used cell-based screens to discover and develop Ceapins, a class of pyrazole amides, that block ATF6α signaling in response to ER stress. Ceapins sensitize cells to ER stress without impacting viability of unstressed cells. Ceapins are highly specific inhibitors of ATF6α signaling, not affecting signaling through the other branches of the UPR, or proteolytic processing of its close homolog ATF6β or SREBP (a cholesterol-regulated transcription factor), both activated by the same proteases. Ceapins are first-in-class inhibitors that can be used to explore both the mechanism of activation of ATF6α and its role in pathological settings. The discovery of Ceapins now enables pharmacological modulation all three UPR branches either singly or in combination.
Sigma-Aldrich
Ceapin-A7, ≥98% (HPLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)