Merck
  • Integrin activation in bovine placentomes and in caruncular epithelial cells isolated from pregnant cows.
Integrin activation in bovine placentomes and in caruncular epithelial cells isolated from pregnant cows.
Biology of reproduction (2008-04-18)
Philip S Bridger, Susanne Haupt, Rudolf Leiser, Gregory A Johnson, Robert C Burghardt, Hans-Rudolf Tinneberg, Christiane Pfarrer
ABSTRACT
In the bovine synepitheliochorial placenta, restricted trophoblast invasion requires complex interactions of integrin receptors with proteins of the extracellular matrix (ECM) and integrin receptors of neighboring cells. Activated integrins assemble to focal adhesions and are linked to the actin cytoskeleton via signaling molecules including alpha-actinin (ACTN), focal adhesion kinase (PTK2 or FAK), phosphotyrosine, and talin (TLN1). Aims of this study were to assess integrin activation and focal adhesion assembly within epithelial cells of bovine placentomes and low-passage (not transformed) placentomal caruncular epithelial cells cultured on dishes coated with ECM proteins. Immunofluorescence analysis was performed to colocalize the signaling molecules ACTN, PTK2, phosphotyrosine, and TLN1 with each other and with beta(1)-integrin (ITGB1) in placentomal cryosections throughout pregnancy and in caruncular epithelial cells in vitro. Antibody specificity was confirmed by Western blot. Cells were cultured on uncoated dishes, and the dishes were coated with fibronectin (FN), laminin (LAMA), and collagen type IV (COL4), thereby statistically assessing cell number and qualitatively assessing the expression pattern of ITGB1, phosphotyrosine, and TLN1. Results demonstrated integrin activation and focal adhesion assembly in the placentome and that low-passage caruncular epithelial cells maintain integrin-associated properties observed in vivo. Expression and/or colocalization of signaling molecules with ITGB1 confirmed, for the first time, integrin activation and participation in "outside-in" and "inside-out" signaling pathways. The prominent role of ECM, and FN in particular, in integrin signaling is supported by the in vitro enhancement of proliferation and focal adhesion expression. Thus, this in vitro model provides excellent potential for further mechanistic studies designed to elucidate feto-maternal interactions in the bovine placentome.
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, FITC conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-Integrin beta1 Antibody, Cytosolic, serum, Chemicon®