Merck
  • A natural human IgM that binds to gangliosides is therapeutic in murine models of amyotrophic lateral sclerosis.
A natural human IgM that binds to gangliosides is therapeutic in murine models of amyotrophic lateral sclerosis.
Disease models & mechanisms (2015-06-03)
Xiaohua Xu, Aleksandar Denic, Luke R Jordan, Nathan J Wittenberg, Arthur E Warrington, Bharath Wootla, Louisa M Papke, Laurie J Zoecklein, Daehan Yoo, Jonah Shaver, Sang-Hyun Oh, Larry R Pease, Moses Rodriguez
ABSTRACT
Amyotrophic lateral sclerosis (ALS) is a devastating, fatal neurological disease that primarily affects spinal cord anterior horn cells and their axons for which there is no treatment. Here we report the use of a recombinant natural human IgM that binds to the surface of neurons and supports neurite extension, rHIgM12, as a therapeutic strategy in murine models of human ALS. A single 200 µg intraperitoneal dose of rHIgM12 increases survival in two independent genetic-based mutant SOD1 mouse strains (SOD1G86R and SOD1G93A) by 8 and 10 days, delays the onset of neurological deficits by 16 days, delays the onset of weight loss by 5 days, and preserves spinal cord axons and anterior horn neurons. Immuno-overlay of thin layer chromatography and surface plasmon resonance show that rHIgM12 binds with high affinity to the complex gangliosides GD1a and GT1b. Addition of rHIgM12 to neurons in culture increases α-tubulin tyrosination levels, suggesting an alteration of microtubule dynamics. We previously reported that a single peripheral dose of rHIgM12 preserved neurological function in a murine model of demyelination with axon loss. Because rHIgM12 improves three different models of neurological disease, we propose that the IgM might act late in the cascade of neuronal stress and/or death by a broad mechanism.
Sigma-Aldrich
Anti-Human IgM (μ-chain specific)-Alkaline Phosphatase, Mouse monoclonal, clone MB-11, purified from hybridoma cell culture
Sigma-Aldrich
Disialoganglioside GD1a from bovine brain, ≥95% (TLC), lyophilized powder
Avanti
Ganglioside GM1 (Ovine Brain), Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
14:0 Liss Rhod PE, Avanti Research - A Croda Brand 810157P, powder
Sigma-Aldrich
Trisialoganglioside-GT1b from bovine brain, lyophilized powder, ≥96% (TLC)
Sigma-Aldrich
Disialoganglioside GD1b from bovine brain, ~95%, lyophilized powder
Sigma-Aldrich
Monosialoganglioside GM1 from bovine brain, lyophilized powder, BioXtra, γ-irradiated, ≥95% (TLC)