Merck
  • Vascular endothelial growth factor receptor 2 (VEGFR-2) plays a key role in vasculogenic mimicry formation, neovascularization and tumor initiation by Glioma stem-like cells.
Vascular endothelial growth factor receptor 2 (VEGFR-2) plays a key role in vasculogenic mimicry formation, neovascularization and tumor initiation by Glioma stem-like cells.
PloS one (2013-03-29)
Xiaohong Yao, Yifang Ping, Ying Liu, Kequiang Chen, Teizo Yoshimura, Mingyong Liu, Wanghua Gong, Chong Chen, Qin Niu, Deyu Guo, Xia Zhang, Ji Ming Wang, Xiuwu Bian
ABSTRACT
Human glioblastomas (GBM) are thought to be initiated by glioma stem-like cells (GSLCs). GSLCs also participate in tumor neovascularization by transdifferentiating into vascular endothelial cells. Here, we report a critical role of GSLCs in the formation of vasculogenic mimicry (VM), which defines channels lined by tumor cells to supply nutrients to early growing tumors and tumor initiation. GSLCs preferentially expressed vascular endothelial growth factor receptor-2 (VEGFR-2) that upon activation by VEGF, mediated chemotaxis, tubule formation and increased expression of critical VM markers by GSLCs. Knockdown of VEGFR-2 in GSLCs by shRNA markedly reduced their capacity of self-renewal, forming tubules, initiating xenograft tumors, promoting vascularization and the establishment of VM. Our study demonstrates VEGFR-2 as an essential molecule to sustain the "stemness" of GSLCs, their capacity to initiate tumor vasculature, and direct initiation of tumor.
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Cy3 conjugate, Species Adsorbed, 1.5 mg/mL, Chemicon®
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, F(ab′)2, FITC conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Anti-Oct-4 Antibody, Chemicon®, from rabbit