Merck
  • Gintonin stimulates gliotransmitter release in cortical primary astrocytes.
Gintonin stimulates gliotransmitter release in cortical primary astrocytes.
Neuroscience letters (2015-07-21)
Hyunsook Kim, Byung-Hwan Lee, Sun-Hye Choi, Hyeon-Joong Kim, Suk-Won Jung, Sung-Hee Hwang, Hyewon Rhim, Hyung-Chun Kim, Ik-Hyun Cho, Seung-Yeol Nah
ABSTRACT
Lysophosphatidic acid (LPA) is a simple and minor phospholipid, but serves as a lipid-derived neurotransmitter via activation of G protein-coupled LPA receptors. Astrocytes abundantly express LPA receptors and contain gliotransmitters that modulate astrocyte-neuron interactions. Gintonin is a novel ginseng-derived G protein-coupled LPA receptor ligand. Gintonin induces [Ca(2+)]i transients in neuronal and non-neuronal cells via activation of LPA receptors, which regulate calcium-dependent ion channels and receptors. A line of evidence shows that neurotransmitter-mediated [Ca(2+)]i elevations in astrocytes are coupled with gliotransmitter release. However, little is known about whether gintonin-mediated [Ca(2+)]i transients are coupled to gliotransmitter release in astrocytes. In the present study, we examined the effects of gintonin on adenosine triphosphate (ATP) and glutamate release in mouse cortical primary astrocytes. Application of gintonin to astrocytes induced [Ca(2+)]i transients in a concentration-dependent and reversible manner. However, ginsenosides, other active ingredients in ginseng, had no effect on [Ca(2+)]i transients. The induction of gintonin-mediated [Ca(2+)]i transients was attenuated/blocked by the LPA1/3 receptor antagonist Ki16425, a phospholipase C inhibitor, an inositol 1,4,5-triphosphate receptor antagonist, and an intracellular Ca(2+) chelator. Gintonin treatment on astrocytes increased ATP and glutamate release in a concentration- and time-dependent manner. BAPTA and Ki16425 attenuated gintonin-mediated ATP and glutamate release in astrocytes. The present study shows that gintonin-mediated [Ca(2+)]i transients are coupled to gliotransmitter release via LPA receptor activation. Finally, gintonin-mediated [Ca(2+)]i transients and gliotransmitter release from astrocytes via LPA receptor activation might explain one mechanism of gintonin-mediated neuromodulation in the central nervous system.
Product Description
SAFC
HEPES
SAFC
HEPES
Avanti
Ganglioside-Total, Avanti Research - A Croda Brand
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Avanti
18:1 Lyso PA, Avanti Research - A Croda Brand
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%