Merck
  • Effect of enzymatic pretreatment of various lignocellulosic substrates on production of phenolic compounds and biomethane potential.
Effect of enzymatic pretreatment of various lignocellulosic substrates on production of phenolic compounds and biomethane potential.
Bioresource technology (2015-06-22)
Michel Schroyen, Han Vervaeren, Hanne Vandepitte, Stijn W H Van Hulle, Katleen Raes
ABSTRACT
Pretreatment of lignocellulosic biomass is necessary to enhance the hydrolysis, which is the rate-limiting step in biogas production. Laccase and versatile peroxidase are enzymes known to degrade lignin. Therefore, the impact of enzymatic pretreatment was studied on a variety of biomass. A significant higher release in total phenolic compounds (TPC) was observed, never reaching the inhibiting values for anaerobic digestion. The initial concentration of TPC was higher in the substrates containing more lignin, miscanthus and willow. The anaerobic digestion of these two substrates resulted in a significant lower biomethane production (68.8-141.7 Nl/kg VS). Other substrates, corn stover, flax, wheat straw and hemp reached higher biomethane potential values (BMP), between 241 and 288 Nl/kg VS. Ensilaged maize reached 449 Nl/kg VS, due to the ensilation process, which can be seen as a biological and acid pretreatment. A significant relation (R(2) = 0.89) was found between lignin content and BMP.
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Sodium malonate dibasic, ≥97.0% (NT)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Sodium malonate dibasic monohydrate, BioXtra
Sigma-Aldrich
Sodium malonate dibasic monohydrate, ≥98% (non-aqueous titration)
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Sigma-Aldrich
2-Hydroxycinnamic acid, predominantly trans, 97%
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Methanol
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%