Merck
  • ApoA-I or ABCA1 expression suppresses fatty acid synthesis by reducing 27-hydroxycholesterol levels.
ApoA-I or ABCA1 expression suppresses fatty acid synthesis by reducing 27-hydroxycholesterol levels.
Biochimie (2014-05-06)
Donglin Ma, Wei Liu, Yutong Wang
ABSTRACT
Abnormal lipid metabolism may contribute to the pathogenesis of non-alcoholic steatohepatitis (NASH). The ATP-binding cassette transporter A1 (ABCA1) protein mediates the transport of cholesterol and phospholipids from cells to apolipoprotein A-I (apoA-I) to generate nascent HDL particles. Previous studies revealed that overexpression of ABCA1 or apoA-I alleviated hepatic lipid levels by modifying lipid transport. Here, we examined the effect of apoA-I and ABCA1 overexpression on genes involved in fatty acid synthesis in QSG-7701 hepatocytes and in mice. Human apoA-I and ABCA1 were overexpressed by transfection in QSG-7701 hepatocytes. Human apoA-I was also overexpressed via an adenoviral vector in C57BL/6J mice fed a methionine choline-deficient diet. Overexpression of either apoA-I or ABCA1 resulted in an increase in cholesterol efflux and a decrease in cellular cholesterol, fatty acids, and triglycerides. Overexpression of these genes also resulted in a reduction in 27-hydroxycholesterol levels and a decrease in the mRNA levels of fatty acid synthase and acetyl-CoA carboxylase 1. Overexpression of apoA-I in mice reduced hepatic lipid levels, 27-hydroxycholesterol levels, and the mRNA levels of fatty acid synthase and acetyl-CoA carboxylase 1. These results suggest that expression of apoA-I or ABCA1 can reduce steatosis by decreasing lipid storage and modifying lipid transport in hepatocytes and may also inhibit fatty acid synthesis by decreasing 27-hydroxycholesterol levels.
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Avanti
22(R)-hydroxycholesterol, Avanti Research - A Croda Brand
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Oleic acid, analytical standard
Supelco
Oleic acid, Selectophore, ≥99%
Avanti
22(R)-hydroxycholesterol-d7, Avanti Research - A Croda Brand