Merck
  • Potential avoidance of adverse analgesic effects using a biologically "smart" hydrogel capable of controlled bupivacaine release.
Potential avoidance of adverse analgesic effects using a biologically "smart" hydrogel capable of controlled bupivacaine release.
Journal of pharmaceutical sciences (2014-10-01)
Francesca Taraballi, Silvia Minardi, Bruna Corradetti, Iman K Yazdi, Marta A Balliano, Jeffrey L Van Eps, Massimo Allegri, Ennio Tasciotti
ABSTRACT
Acute pain remains a tremendous clinical and economic burden, as its prevalence and common narcotic-based treatments are associated with poorer outcomes and higher costs. Multimodal analgesia portends great therapeutic promise, but rarely allows opioid sparing, and new alternatives are necessary. Microparticles (MPs) composed of biodegradable polymers [e.g., poly(lactic-co-glycolic acid) or PLGA] have been applied for controlled drug release and acute pain treatment research. However, foreign particles' presence within inflamed tissue may affect the drug release or targeting, and/or cause a secondary inflammatory reaction. We examined how small alterations in the particulate nature of MPs affect both their uptake into and subsequent activation of macrophages. MPs composed of PLGA and chitosan (PLGA-Chi) loaded with bupivacaine (BP) were engineered at different sizes and their opsonization by J774 macrophages was assessed. Uptake of PLGA-Chi by macrophages was found to be size dependent, but they were not cytotoxic or proinflammatory in effect. Moreover, encapsulation of MPs in a thermoresponsive loading gel (pluronic F-127) effectively prevented opsonization. Finally, MPs displayed sustained, tunable release of BP up to 7 days. These results demonstrate our ability to develop a drug delivery system capable of controlled release of local anesthetics to treat acute/subacute pain while concurrently avoiding enhanced inflammation.
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Supelco
Bupivacaine hydrochloride monohydrate, analytical standard, for drug analysis
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Rhodamine B
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Supelco
Rhodamine B, analytical standard
Supelco
Bupivacaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Bupivacaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DAPI, for nucleic acid staining