Merck
  • Microbial synthesis of polyhydroxyalkanoate using seaweed-derived crude levulinic acid as co-nutrient.
Microbial synthesis of polyhydroxyalkanoate using seaweed-derived crude levulinic acid as co-nutrient.
International journal of biological macromolecules (2014-09-07)
Anupam Bera, Sonam Dubey, Khushbu Bhayani, Dibyendu Mondal, Sandhya Mishra, Pushpito K Ghosh
ABSTRACT
Production of polyhydroxyalkanoates (PHAs) from Jatropha biodiesel residues, namely crude glycerol and oil cake hydrolysate, has been reported previously. Halomonas hydrothermalis (MTCC accession no. 5445; NCBI Genbank accession no. GU938192), a wild marine strain, was used in the bio-synthesis. The present study was initiated to vary the properties of the polymer. Seaweed-derived crude levulinic acid (SDCLA), containing formic acid, residual sugars and dissolved minerals additionally, was proposed as co-feed along with the biodiesel residues. Experiments were conducted at 100mL scale in batch process. Whereas the PHA yield was only 0.40 ± 0.01 g when only biodiesel residues were employed, it rose to 1.07 ± 0.02 g in presence of 0.35% (w/v) of SDCLA. The corresponding carbon utilisation efficiencies were 29.3% and 57.5%, respectively. 3-Hydroxy valerate incorporation in the PHA was pronounced in presence of SDCLA, with associated changes in polymer properties. The microbial synthesis fared poorly when SDCLA was substituted with pure levulinic acid. Thus, Halomonas hydrothermalis had a poor response to levulinic acid, as such, and other constituents present in SDCLA appear to have played a vital role in bacterial cell division and accumulation of PHA. Biodegradability tests in moist soil were also conducted as part of the study. Marine microalgal cultivation for biodiesel and seaweed cultivation for fuels may help generate biodiesel residues and crude levulinic acid in proximity, which would open up the possibility of large scale PHA manufacture in efficient and practical manner in the future through the methodology of the present study.
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Methanol
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%