Merck
  • Fluoxetine suppresses AMP-activated protein kinase signaling pathway to promote hepatic lipid accumulation in primary mouse hepatocytes.
Fluoxetine suppresses AMP-activated protein kinase signaling pathway to promote hepatic lipid accumulation in primary mouse hepatocytes.
The international journal of biochemistry & cell biology (2014-08-08)
Jing Xiong, Huan Yang, Lili Wu, Wei Shang, Enfang Shan, Wei Liu, Gang Hu, Tao Xi, Jian Yang
ABSTRACT
In the previous study, we demonstrated that fluoxetine (FLX) regulated lipogenic and lipolytic genes to promote hepatic lipid accumulation. On this basis, underlying mechanisms were investigated by focusing on the intracellular signaling transduction in the present study using primary mouse hepatocytes. The expression of lipogenesis- and lipolysis-related genes was evaluated with the application of specific activators and inhibitors. Activation status of respective signaling pathway and the lipid accumulation in hepatocytes were analyzed. We provided evidence that AMP-activated protein kinase (AMPK) activator AICAR (5-aminoimidazole-4-carboxamide-1-β-D-ribofuranoside) significantly suppressed the increased expression of representative lipogenesis-related genes, acetyl-CoA carboxylase (ACC) and fatty acid synthase (FAS) by FLX, while increased the repressed expression of lipolysis-related genes, carboxylesterases. In the meanwhile, FLX regulated the above genes in the same way as AMPK inhibitor Compound C did. Furthermore, AICAR inhibited the proteolytic activation of SREBP1c induced by FLX, resulting in the decreased level of nuclear SREBP1c. Further studies demonstrated that FLX significantly suppressed the phosphorylation of AMPK and subsequent phosphorylation of ACC, following the inhibited phosphorylation and nuclear export of liver kinase B1 (LKB1). As a functional analysis, FLX-induced lipid accumulation in hepatocytes was repeatedly abolished by AICAR. In conclusion, FLX-induced hepatic lipid accumulation is mediated by the suppression of AMPK signaling pathway. The findings not only provide new insight into the understanding of the mechanisms for selective serotonin reuptake inhibitors-mediated dyslipidemia effects, but also suggest a novel therapeutic target to interfere.
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Dacarbazine impurity B, European Pharmacopoeia (EP) Reference Standard