Merck
  • Tyrosine kinase inhibitor tyrphostin AG490 retards chronic joint inflammation in mice.
Tyrosine kinase inhibitor tyrphostin AG490 retards chronic joint inflammation in mice.
Inflammation (2014-01-30)
Valeriya Gyurkovska, Tsvetanka Stefanova, Petya Dimitrova, Svetla Danova, Rositsa Tropcheva, Nina Ivanovska
ABSTRACT
Tyrphostin AG490 is a Janus kinase (JAK) 2 inhibitor that is clinically used as an anticancer agent and is also effective in various models of inflammatory and autoimmune diseases. In this study, we examined the effects of tyrphostin AG490 on the development of collagenase-induced osteoarthritis (CIOA). Our results showed that tyrphostin-ameliorated cartilage and bone destructions. This effect was associated with decreased expression of signal transducers and activators of transcription 3 (STAT3), phosphorylated JAK2, Dickkopf homolog 1, and receptor activator of nuclear factor κB ligand (RANKL) in the joints of arthritic mice. Tyrphostin AG490 suppressed STAT3 phosphorylation and the expression of tumor necrosis factor-related apoptosis-inducing ligand and RANKL by synovial fluid cells. The drug inhibited RANKL-induced osteoclast differentiation in vitro. Molecules, such as tyrphostin AG490 that limit bone erosion and influence osteoclast generation, might have therapeutic utility in joint degenerative disorders.
Product Description
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material