Merck
  • A simple method for the small scale synthesis and solid-phase extraction purification of steroid sulfates.
A simple method for the small scale synthesis and solid-phase extraction purification of steroid sulfates.
Steroids (2014-10-07)
Christopher C Waller, Malcolm D McLeod
ABSTRACT
Steroid sulfates are a major class of steroid metabolite that are of growing importance in fields such as anti-doping analysis, the detection of residues in agricultural produce or medicine. Despite this, many steroid sulfate reference materials may have limited or no availability hampering the development of analytical methods. We report simple protocols for the rapid synthesis and purification of steroid sulfates that are suitable for adoption by analytical laboratories. Central to this approach is the use of solid-phase extraction (SPE) for purification, a technique routinely used for sample preparation in analytical laboratories around the world. The sulfate conjugates of sixteen steroid compounds encompassing a wide range of steroid substitution patterns and configurations are prepared, including the previously unreported sulfate conjugates of the designer steroids furazadrol (17β-hydroxyandrostan[2,3-d]isoxazole), isofurazadrol (17β-hydroxyandrostan[3,2-c]isoxazole) and trenazone (17β-hydroxyestra-4,9-dien-3-one). Structural characterization data, together with NMR and mass spectra are reported for all steroid sulfates, often for the first time. The scope of this approach for small scale synthesis is highlighted by the sulfation of 1μg of testosterone (17β-hydroxyandrost-4-en-3-one) as monitored by liquid chromatography-mass spectrometry (LCMS).
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Methanol
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
1,4-Dioxane, ReagentPlus®, ≥99%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Potassium permanganate, ≤150 μm particle size, 97%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
1,4-Dioxane, analytical standard
Supelco
Methanol, analytical standard
Supelco
Androsterone, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%
Supelco
1,4-Dioxane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
trans-Androsterone