Merck
  • Altered expression of dopamine receptors in cholinergic motoneurons of the hypoglossal nucleus in a 6-OHDA-induced Parkinson's disease rat model.
Altered expression of dopamine receptors in cholinergic motoneurons of the hypoglossal nucleus in a 6-OHDA-induced Parkinson's disease rat model.
Biochemical and biophysical research communications (2014-08-31)
Li Zhou, Zhi-Yong Wang, Hui Lian, Hai-Yan Song, Yi-Min Zhang, Xiao-Li Zhang, Rui-Fang Fan, Li-Fei Zheng, Jin-Xia Zhu
ABSTRACT
Parkinson's disease (PD) is a common neurodegenerative disorder that is often associated with weak tongue motility. However, the link between the degenerated dopaminergic neurons in the substantia nigra (SN) and lingual dysfunction remains unclear. In the present study, we investigated the localization of dopamine receptor 1 (D1) and dopamine receptor 2 (D2) and alternations in their expression in cholinergic motoneurons of the hypoglossal nucleus (HN) using double-label immunofluorescence, Western blotting and semi-quantitative reverse transcription and polymerase chain reaction (SqRT-PCR) in rats that received microinjections of 6-hydroxydopamine bilaterally into the SN (6-OHDA rats). The results revealed that a large population of choline acetyltransferase immunoreactive (ChAT-IR) neurons was distributed throughout HN and that almost all of the ChAT-IR motoneurons were also D1-IR and D2-IR. Several tyrosine hydroxylase (TH)-IR profiles were observed in a nonuniform pattern near the ChAT-IR, D1-IR or D2-IR somas, suggesting potent dopaminergic innervation. In the 6-OHDA rats, TH immunoreactivity in the SN was significantly decreased, but food residue was increased and treadmill occupancy time was shortened. In the HN, protein expression of TH and D2 was increased, whereas that of ChAT and D1 was decreased. A similar pattern was observed in mRNA levels. The present study suggests that dopamine may modulate the activity of cholinergic neurons via binding with D1 and D2 in the HN. Changes in the expression of ChAT, TH, D1 and D2 in the HN of 6-OHDA rats might be associated with the impaired tongue motility in PD. These findings should be further investigated.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis