Merck
  • Release of betaine and dexpanthenol from vitamin E modified silicone-hydrogel contact lenses.
Release of betaine and dexpanthenol from vitamin E modified silicone-hydrogel contact lenses.
Current eye research (2014-05-17)
Kuan-Hui Hsu, Percy Lazon de la Jara, Amali Ariyavidana, Jason Watling, Brien Holden, Qian Garrett, Anuj Chauhan
ABSTRACT
To develop a contact lens system that will control the release of an osmoprotectant and a moisturizing agent with the aim to reduce symptoms of ocular dryness. Profiles of the release of osmoprotectant betaine and moisturizing agent dexpanthenol from senofilcon A and narafilcon B contact lenses were determined in vitro under sink conditions. Both types of lenses were also infused with vitamin E to increase the duration of drug release due to the formation of the vitamin E diffusion barriers in the lenses. The release profiles from vitamin E-infused lenses were compared with those from the control lenses. Both dexpanthenol and betaine are released from commercial silicone hydrogel lenses for only about 10 min. Vitamin E loadings into contact lenses at about 20-23% can increase the release times to about 10 h, which is about 60 times larger compared to the control unmodified lenses. Vitamin E-loaded silicone hydrogel contact lenses released betaine and dexpanthenol in a controlled fashion.
Product Description
Dexpanthenol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dexpanthenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)