Merck
  • Synthesis and cellular internalization of spindle hematite/polymer hybrid nanoparticles.
Synthesis and cellular internalization of spindle hematite/polymer hybrid nanoparticles.
ACS applied materials & interfaces (2015-02-19)
Jing Wang, Wen Zhu, Lixin Liu, Yongming Chen, Chun Wang
ABSTRACT
Nonspherical spindle-shaped hematite/polymer hybrid nanoparticles (SPNPs) were synthesized via surface-initiated atom transfer radical polymerization (SI-ATRP). The long axis of the SPNPs was 370 ± 65 nm, and the short axis was 80 ± 15 nm with an aspect ratio of 4.6-4.7. The SPNPs were characterized by transmission electron microscopy (TEM), Fourier transform infrared spectroscopy (FT-IR), and X-ray photoelectron spectroscopy (XPS). Thermogravimetric analysis (TGA) was used to estimate the content of grafted polymer. Light-scattering measurement was used to detect the particle size distribution of SPNPs in water and in cell culture medium. HeLa cells internalized the SPNPs within 1 h, and the uptake reached equilibrium in 8 h. These observations contribute to better understanding of the interactions between nonspherical nanoparticles and cells, which may have implication for designing drug delivery vehicles.
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Ethyl α-bromoisobutyrate, 98%
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Supelco
Rhodamine B, analytical standard
Sigma-Aldrich
Rhodamine B
Sigma-Aldrich
Copper(I) bromide, 99.999% trace metals basis
Sigma-Aldrich
Copper(I) bromide, AnhydroBeads, -10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Copper(I) bromide, 98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
α-Bromoisobutyryl bromide, 98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%