Merck
  • Polycaprolactone/oligomer compound scaffolds for cardiac tissue engineering.
Polycaprolactone/oligomer compound scaffolds for cardiac tissue engineering.
Journal of biomedical materials research. Part A (2013-11-30)
Chaganti Srinivasa Reddy, Jayarama Reddy Venugopal, Seeram Ramakrishna, Eyal Zussman
ABSTRACT
Polycaprolactone (PCL), a synthetic biocompatible and biodegradable polymer generally used as a scaffold material for tissue engineering applications. The high stiffness and hydrophobicity of the PCL fiber mesh does not provide significant cell attachment and proliferation in cardiac tissue engineering. Towards this goal, the study focused on a compound of PCL and oligomer hydrogel [Bisphenol A ethoxylated dimethacrylate (BPAEDMA)] processed into electrospun nanofibrous scaffolds. The composition, morphology and mechanical properties of the compound scaffolds, composed of varying ratios of PCL and hydrogel were characterized by scanning electron microscopy, infrared spectroscopy and dynamic mechanical analyzer. The elastic modulus of PCL/BPAEDMA nanofibrous scaffolds was shown to be varying the BPAEDMA weight fraction and was decreased by increasing the BPAEDMA weight fraction. Compound fiber meshes containing 75 wt % BPAEDMA oligomer hydrogel exhibited lower modulus (3.55 MPa) and contact angle of 25(o) . Rabbit cardiac cells cultured for 10 days on these PCL/BPAEDMA compound nanofibrous scaffolds remained viable and expressed cardiac troponin and alpha-actinin proteins for the normal functioning of myocardium. Cell adhesion and proliferations were significantly increased on compound fiber meshes containing 75 wt % BPAEDMA, when compared with other nanofibrous scaffolds. The results observed that the produced PCL/BPAEDMA compound nanofibrous scaffolds promote cell adhesion, proliferation and normal functioning of cardiac cells to clinically beneficial levels, relevant for cardiac tissue engineering.
Sigma-Aldrich
Magnesium sulfate solution, BioUltra
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Hexamethyldisilazane, Wacker Chemie AG, ≥97.0% (GC)
Supelco
Hexamethyldisilazane, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Magnesium sulfate solution, 1.00 M±0.04 M
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture