Merck
  • Association of folic acid supplementation during pregnancy and infant bronchiolitis.
Association of folic acid supplementation during pregnancy and infant bronchiolitis.
American journal of epidemiology (2014-03-29)
Sreenivas P Veeranki, Tebeb Gebretsadik, Stacy L Dorris, Edward F Mitchel, Tina V Hartert, William O Cooper, Frances A Tylavsky, William Dupont, Terryl J Hartman, Kecia N Carroll
ABSTRACT
Viral bronchiolitis affects 20%-30% of infants; because there is no known effective treatment, it is important to identify risk factors that contribute to its pathogenesis. Although adequate folate intake during the periconceptional period prevents neural tube defects, animal data suggest that higher supplementation may be a risk factor for child respiratory diseases. Using a population-based retrospective cohort of 167,333 women and infants, born in 1995-2007 and enrolled in the Tennessee Medicaid program, we investigated the association between the filling of folic acid-containing prescriptions and infant bronchiolitis. We categorized women into the following 4 groups in relation to the first trimester: "none" (no prescription filled), "first trimester only," "after first trimester," and "both" (prescriptions filled both during and after the first trimester). Overall, 21% of infants had a bronchiolitis diagnosis, and 5% were hospitalized. Most women filled their first prescriptions after the fifth to sixth weeks of pregnancy, and most prescriptions contained 1,000 µg of folic acid. Compared with infants born to women in the "none" group, infants born to women in the "first trimester only" group had higher relative odds of bronchiolitis diagnosis (adjusted odds ratio = 1.17, 95% confidence interval: 1.11, 1.22) and greater severity (adjusted odds ratio = 1.16, 95% confidence interval: 1.11, 1.22). This study's findings contribute to an understanding of the implications of prenatal nutritional supplement recommendations for infant bronchiolitis.
Product Description
Folic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, meets USP testing specifications
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard