Merck
  • Inhibition of dipeptidyl peptidase IV and xanthine oxidase by amino acids and dipeptides.
Inhibition of dipeptidyl peptidase IV and xanthine oxidase by amino acids and dipeptides.
Food chemistry (2013-06-19)
Alice B Nongonierma, Catherine Mooney, Denis C Shields, Richard J Fitzgerald
ABSTRACT
Xanthine oxidase (XO) and dipeptidyl peptidase IV (DPP-IV) inhibition by amino acids and dipeptides was studied. Trp and Trp-containing dipeptides (Arg-Trp, Trp-Val, Val-Trp, Lys-Trp and Ile-Trp) inhibited XO. Three amino acids (Met, Leu and Trp) and eight dipeptides (Phe-Leu, Trp-Val, His-Leu, Glu-Lys, Ala-Leu, Val-Ala, Ser-Leu and Gly-Leu) inhibited DPP-IV. Trp and Trp-Val were multifunctional inhibitors of XO and DPP-IV. Lineweaver and Burk analysis showed that Trp was a non-competitive inhibitor of XO and a competitive inhibitor of DPP-IV. Molecular docking with Autodock Vina was used to better understand the interaction of the peptides with the active site of the enzyme. Because of the non-competitive inhibition observed, docking of Trp-Val to the secondary binding sites of XO and DPP-IV is required. Trp-Val was predicted to be intestinally neutral (between 25% and 75% peptide remaining after 60 min simulated intestinal digestion). These results are of significance for the reduction of reactive oxygen species (ROS) and the increase of the half-life of incretins by food-derived peptides.
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade IV, ammonium sulfate suspension, ≥0.1 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, lyophilized powder, 0.4-1.0 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade III, ammonium sulfate suspension, ≥0.8 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade I, ammonium sulfate suspension, ≥0.4 units/mg protein
Sigma-Aldrich
Xanthine Oxidase microbial, lyophilized powder, ≥7 units/mg solid