Merck
  • Recombinant human thrombomodulin suppresses experimental abdominal aortic aneurysms induced by calcium chloride in mice.
Recombinant human thrombomodulin suppresses experimental abdominal aortic aneurysms induced by calcium chloride in mice.
Annals of surgery (2013-01-09)
Chao-Han Lai, Guey-Yueh Shi, Fang-Tzu Lee, Cheng-Hsiang Kuo, Tsung-Lin Cheng, Bi-Ing Chang, Chih-Yuan Ma, Fu-Chih Hsu, Yu-Jen Yang, Hua-Lin Wu
ABSTRACT
To investigate whether recombinant thrombomodulin containing all the extracellular domains (rTMD123) has therapeutic potential against aneurysm development. The pathogenesis of abdominal aortic aneurysm (AAA) is characterized by chronic inflammation and proteolytic degradation of extracellular matrix. Thrombomodulin, a transmembrane glycoprotein, exerts anti-inflammatory activities such as inhibition of cytokine production and sequestration of proinflammatory high-mobility group box 1 (HMGB1) to prevent it from engaging the receptor for advanced glycation end product (RAGE) that may sustain inflammation and tissue damage. The in vivo effects of treatment and posttreatment with rTMD123 on aortic dilatation were measured using the CaCl2-induced AAA model in mice. Characterization of the CaCl2-induced model revealed that HMGB1 and RAGE, both localized mainly to macrophages, were persistently upregulated during a 28-day period of AAA development. In vitro, rTMD123-HMGB1 interaction prevented HMGB1 binding to macrophages, thereby prohibiting activation of HMGB1-RAGE signaling in macrophages. In vivo, short-term treatment with rTMD123 upon AAA induction suppressed the levels of proinflammatory cytokines, HMGB1, and RAGE in the aortic tissue; reduced the infiltrating macrophage number; and finally attenuated matrix metalloproteinase production, extracellular matrix destruction, and AAA formation without disturbing vascular calcification. Consistently, posttreatment with rTMD123 seven days after AAA induction alleviated vascular inflammation and retarded AAA progression. These data suggest that rTMD123 confers protection against AAA development. The mechanism of action may be associated with reduction of proinflammatory mediators, blockade of macrophage recruitment, and suppression of HMGB1-RAGE signaling involved in aneurysm formation and downstream macrophage activation.
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride solution, BioUltra, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, ≥99.0%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%